Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Tubacin
GREENER_CHOICESDP

Catalog No. 34021 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-021 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 34-021 Supplier R&D Systems™ Supplier No. 3402/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. HDAC6 inhibitor; inhibits α-tubulin deacetylation

Selective inhibitor of HDAC6; inhibits the second deacetylase domain (DD2). Does not inhibit HDAC6 histone deacetylase activity; reversibly inhibits α-tubulin deacetylation. Increases α-tubulin acetylation levels with no effect on histone acetylation or cell cycle progression.

Chemical Identifiers

CAS 1350555-93-9
Molecular Formula C41H43N3O7S

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 721.86
Purity >98%
Molecular Weight (g/mol) 721.869
Chemical Name or Material Tubacin
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.