Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBAL3 Monoclonal Antibody (OTI4C5), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA503909

Encompass_Preferred

 View more versions of this product

Catalog No. 50-167-4686


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha chain.
Specifications

Specifications

TUBAL3
Monoclonal
0.55 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
A6NHL2, Q3UX10
TUBAL3
Human recombit protein fragment corresponding to amino acids 150-446 of human TUBAL3 produced in E.coli.
100 μL
Primary
Canine, Human, Mouse, Rat
Antibody
IgG2b
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI4C5
Unconjugated
TUBAL3
tubulin alpha like 3
Mouse
Affinity Chromatography
RUO
238463, 291287, 487148, 79861
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.