Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TUBAL3 Monoclonal Antibody (OTI5F10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503911S


Description
TUBAL3 Monoclonal antibody specifically detects TUBAL3 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
TUBAL3 | |
Monoclonal | |
1.1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q14722 | |
Mouse | |
Affinity chromatography | |
RUO | |
7881 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Immunocytochemistry, Western Blot | |
OTI5F10 | |
Unconjugated | |
KCNAB1 | |
TUBAL3; Tubulin alpha chain-like 3; tubulin alpha like 3; tubulin, alpha-like 3 | |
Full length human recombinant protein of human KCNAB1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction