Learn More
TUBAL3 Mouse anti-Human, Clone: OTI4D8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503907

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha chain.Specifications
TUBAL3 | |
Monoclonal | |
Unconjugated | |
TUBAL3 | |
tubulin alpha like 3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
79861 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI4D8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
A6NHL2 | |
TUBAL3 | |
Human recombit protein fragment corresponding to amino acids 150-446 of human TUBAL3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.