Learn More
TUBB1 Mouse anti-Human, Clone: OTI1B9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF506654

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TUBB1 encodes a member of the beta tubulin protein family. Beta tubulins are one of two core protein families (alpha and beta tubulins) that heterodimerize and assemble to form microtubules. This protein is specifically expressed in platelets and megakaryocytes and may be involved in proplatelet production and platelet release. A mutations in this gene is associated with autosomal domit macrothrombocytopenia. Two pseudogenes of this gene are found on chromosome Y.Specifications
TUBB1 | |
Monoclonal | |
Unconjugated | |
TUBB1 | |
tubulin beta 1 class VI | |
Mouse | |
Affinity Chromatography | |
RUO | |
81027 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1B9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9H4B7 | |
TUBB1 | |
Full length human recombit protein of human TUBB1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.