Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUBB2A Monoclonal Antibody (6A10B2)
GREENER_CHOICE

Catalog No. PIMA531941
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531941 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531941 Supplier Invitrogen™ Supplier No. MA531941
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31941 has been tested in indirect ELISA.

TUBB2A belongs to the tubulin family. Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha-chain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUBB2A
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6A10B2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene TUBB2A
Gene Accession No. P85108, Q13885, Q7TMM9
Gene Alias CDCBM5; class IIa beta-tubulin; dJ40E16.7; M(beta)2; TUBB; Tubb2; TUBB2A; tubb2a {ECO:0000250; tubulin beta 2A class IIa; Tubulin beta class IIa; Tubulin beta-2A chain; tubulin, beta 2A class IIA; tubulin, beta polypeptide; tubulin, beta polypeptide 2; UniProtKB:Q7TMM9}
Gene Symbols TUBB2A
Host Species Mouse
Immunogen Purified recombinant fragment of human TUBB2A (AA: 25-187) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22151, 498736, 7280
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.