Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TUBB2A Polyclonal Antibody

Supplier: Thermo Scientific PA5112780
Description
Specifications
| TUBB2A | |
| Polyclonal | |
| Unconjugated | |
| TUBB2A | |
| CDCBM5; class IIa beta-tubulin; dJ40E16.7; M(beta)2; TUBB; Tubb2; TUBB2A; tubb2a {ECO:0000250; tubulin beta 2A class IIa; Tubulin beta class IIa; Tubulin beta-2A chain; tubulin, beta 2A class IIA; tubulin, beta polypeptide; tubulin, beta polypeptide 2; UniProtKB:Q7TMM9} | |
| Rabbit | |
| Protein G | |
| RUO | |
| 7280 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 3.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q13885 | |
| TUBB2A | |
| Recombinant Human Tubulin beta-2A chain protein (1-445AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction