Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUBB2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107045
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107045 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107045 Supplier Invitrogen™ Supplier No. PA5107045
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Human: 100%; Mouse: 100%; Rat: 100%; Sheep: 100%.

TUBB2A belongs to the tubulin family. Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha-chain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUBB2A
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene TUBB2A
Gene Accession No. Q13885
Gene Alias CDCBM5; class IIa beta-tubulin; dJ40E16.7; M(beta)2; TUBB; Tubb2; TUBB2A; tubb2a {ECO:0000250; tubulin beta 2A class IIa; Tubulin beta class IIa; Tubulin beta-2A chain; tubulin, beta 2A class IIA; tubulin, beta polypeptide; tubulin, beta polypeptide 2; UniProtKB:Q7TMM9}
Gene Symbols TUBB2A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TUBB2A (aa 254-304).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7280
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.