Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBB2B Mouse anti-Human, Clone: OTI5D3, TrueMAB™, Origene
SDP

Catalog No. 502242430
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-224-2430 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-224-2430 Supplier OriGene Supplier No. TA506601S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TUBB2B Monoclonal antibody specifically detects TUBB2B in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen TUBB2B
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5D3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TUBB2B
Gene Accession No. Q9BVA1
Gene Alias 2410129E14Rik; bA506K6.1; beta-tubulin T beta15 (aa 1-445); brdp; class II beta-tubulin isotype; class IIb beta-tubulin; PMGYSA; RGD1309427; T beta-15; Tubb2; Tubb2a; TUBB2B; tubulin beta 2B class IIb; tubulin beta-2B chain; tubulin, beta 2; tubulin, beta 2B class IIb; tubulin, beta polypeptide paralog
Host Species Mouse
Immunogen Full length human recombinant protein of human TUBB2B produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347733
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.