Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBB2B Mouse anti-Human, Clone: OTI6D11, lyophilized, TrueMAB™
SDP

Catalog No. 501664362
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-4362 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-4362 Supplier OriGene Supplier No. CF506600
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

MA1-82191 recognizes class III beta-tubulin whixh is reported to be restricted to neuronal tissue. Clone TU-20 has been used to investigate tumors of neuronal origin including neuroblastoma and ganglioneuroma. Class III beta tubulin has not been detected in tumors of non neuronal origin.

Specifications

Antigen TUBB2B
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone OTI6D11
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene TUBB2B
Gene Accession No. Q9BVA1
Gene Alias CDCBM7, PMGYSA, bA506K6.1
Gene Symbols TUBB2B
Host Species Mouse
Immunogen Full length human recombit protein of human TUBB2B produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347733
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.