Learn More
TUBB2B, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA560448
Description
MA1-82191 recognizes class III beta-tubulin whixh is reported to be restricted to neuronal tissue. Clone TU-20 has been used to investigate tumors of neuronal origin including neuroblastoma and ganglioneuroma. Class III beta tubulin has not been detected in tumors of non neuronal origin.
Immunogen sequence: SILPSPKVSD TVVEPYNATL SVHQLIENAD ETFCIDNEAL YDICSHighest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
Specifications
TUBB2B | |
Polyclonal | |
Unconjugated | |
TUBB2B | |
Tubulin beta-2B chain, TUBB2B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
291081, 347733, 73710 | |
Store at 4°C short term. Store at −20°C long term. Avoid freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BVA1, Q9CWF2, Q3KRE8 | |
TUBB2B | |
Recombinant protein corresponding to Human TUBB2B | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only