Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBB3 Mouse anti-Human, Clone: 6F12, Bioss
SDP

Catalog No. 501979934
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9934 Supplier Bioss Supplier No. BSM33177M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The betaIII-tubulin isoform is present dominantly in cells of neuronal origin and it is one of the earliest markers of neuronal differentiation. It is regarded as a specific probe for the cells of neuronal origin as well as for the tumours originating from these cells. The betaIII-tubulin is most abundant in cells of neuronal origin, but was also detected in Sertoli cells of the testis and transiently in non-neuronal embryonic tissues.

Specifications

Antigen TUBB3
Applications Western Blot
Classification Monoclonal
Clone 6F12
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TUBB3
Gene Accession No. Q13509
Gene Alias 3200002H15Rik; beta-4; beta-4-tubulin; betaIII-tubulin; Beta-tubulin class-III; BOS_17166; CDCBM; CDCBM1; CFEOM3; CFEOM3A; class III beta-tubulin; FEOM3; I79_000194; LOC100341581; M(beta)3; M(beta)6; MC1R; Neuron-specific class III beta-tubulin; TUBB; TUBB3; TUBB4; tubulin beta 3 class III; Tubulin beta-3 chain; Tubulin beta-4 chain; Tubulin beta-III; tubulin, beta 3 class III; tubulin, beta, 3; tubulin, beta, 4; tubulin, beta, 4-like
Gene Symbols TUBB3
Host Species Mouse
Immunogen KLH conjugated synthetic peptide derived from human TUBB3.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10381
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.