Learn More
TUBB3 Mouse anti-Human, Mouse, Rat, Clone: 1F8G10, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 663751IG150UL

Description
This antibody is specific to TUBB3 but not cross-react with other tubulin isoforms.
The betaIII-tubulin isoform is present domitly in cells of neuronal origin and it is one of the earliest markers of neuronal differentiation. It is regarded as a specific probe for the cells of neuronal origin as well as for the tumours originating from these cells. The betaIII-tubulin is most abundant in cells of neuronal origin, but was also detected in Sertoli cells of the testis and transiently in non-neuronal embryonic tissues.Specifications
TUBB3 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13509, Q4QRB4, Q9ERD7 | |
TUBB3 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1F8G10 | |
Unconjugated | |
TUBB3 | |
beta 4, beta tubulin, MC1R, TUBB, TUBB3, TUBB3-specific, Tubulin beta 3 chain, Tubulin beta III, tubulin, beta 3, Tuj1 | |
Mouse | |
Protein G | |
RUO | |
10381, 22152, 246118 | |
-20°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.