Learn More
TUBB4 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI5C1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503129

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TUBB4A (beta-4 tubulin) encodes a member of the beta tubulin family. Beta tubulins are one of two core protein families (alpha and beta tubulins) that heterodimerize and assemble to form microtubules. Mutations in this gene cause hypomyelinating leukodystrophy-6 and autosomal domit torsion dystonia-4. Alternate splicing results in multiple transcript variants encoding different isoforms. A pseudogene of this gene is found on chromosome X.Specifications
TUBB4 | |
Monoclonal | |
Unconjugated | |
TUBB4A | |
DYT4, TUBB4, beta-5 | |
Mouse | |
Affinity Chromatography | |
RUO | |
100855815, 10382, 22153, 29213 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI5C1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P04350, Q9D6F9 | |
TUBB4A | |
Full length human recombit protein of human TUBB4 produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.