Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBB4 Mouse anti-Canine, Human, Mouse, Rat, HRP, Clone: OTI5C1, liquid, TrueMAB™
SDP

Catalog No. 501674097
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-4097 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-4097 Supplier OriGene Supplier No. TA503129BM
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TUBB4A (beta-4 tubulin) encodes a member of the beta tubulin family. Beta tubulins are one of two core protein families (alpha and beta tubulins) that heterodimerize and assemble to form microtubules. Mutations in this gene cause hypomyelinating leukodystrophy-6 and autosomal domit torsion dystonia-4. Alternate splicing results in multiple transcript variants encoding different isoforms. A pseudogene of this gene is found on chromosome X.

Specifications

Antigen TUBB4
Applications Western Blot
Classification Monoclonal
Clone OTI5C1
Conjugate HRP
Formulation PBS with 0.1% BSA, 50% glycerol and 0.02% sodium azide
Gene TUBB4A
Gene Accession No. P04350, Q9D6F9
Gene Alias DYT4, TUBB4, beta-5
Gene Symbols Tubb4a
Host Species Mouse
Immunogen Full length human recombit protein of human TUBB4 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100855815, 10382, 22153, 29213
Target Species Canine, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.