Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBB4B Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: DOF-20, Abnova™

Catalog No. 89336141 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-336-141 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-336-141 Supplier Abnova Corporation Supplier No. MAB19616
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

TUBB4B Monoclonal antibody specifically detects TUBB4B in Human, Mouse, Rat samples. It is validated for IP, ICC, WB-Ce, IHC-P, IF, Flow Cyt.

Specifications

Antigen TUBB4B
Applications Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone DOF-20
Conjugate Unconjugated
Description tubulin, beta 4B class IVb
Gene TUBB4B
Gene Alias RP13-122B23.2, Beta2, TUBB2, TUBB2C
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human TUBB4B
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 10383
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.