Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TUBB4Q Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548754
Description
TUBB4Q Polyclonal Antibody for Western Blot

Specifications
TUBB4Q | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
0 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
56604 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
Unconjugated | |
TUBB7P | |
TUBB7P | |
KLH conjugated synthetic peptide between 64-93 amino acids from the N-terminal region of human TUBB4Q. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction