Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUBB6 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA548312

Catalog No. PA548312


Only null left
Add to Cart

Description

Description

Predicted to react with bovine, monkey and mouse based on sequence homology.

Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha chain.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TUBB6
Polyclonal
Unconjugated
TUBB6
2310057H16Rik; BB220206; class V beta-tubulin; HsT1601; TUBB-5; Tubb6; tubulin beta; tubulin beta 6 class V; tubulin beta class V; tubulin beta MGC4083; tubulin beta-6 chain; tubulin, beta 6 class V
Rabbit
Antigen affinity chromatography, Protein A
RUO
84617
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q9BUF5
TUBB6
KLH conjugated synthetic peptide between 139-166 amino acids from the Central region of human TUBB6.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.