Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUBB6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119484
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119484 200 μL
PIPA5122206 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119484 Supplier Invitrogen™ Supplier No. PA5119484
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TUBB6 Polyclonal Antibody for Western Blot, IHC (P)

Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha chain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUBB6
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene TUBB6
Gene Accession No. Q922F4, Q9BUF5
Gene Alias 2310057H16Rik; BB220206; class V beta-tubulin; HsT1601; TUBB-5; Tubb6; tubulin beta; tubulin beta 6 class V; tubulin beta class V; tubulin beta MGC4083; tubulin beta-6 chain; tubulin, beta 6 class V
Gene Symbols TUBB6
Host Species Rabbit
Immunogen Recombinant protein Tubulin Beta 6. The antigen corresponds to amino acid range 194-421 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 307351, 67951, 84617
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.