Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUBB8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597035
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597035 Supplier Invitrogen™ Supplier No. PA597035
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-251MG, Jurkat, K-562, Mouse brain; Cellular Location: Cytoplasm, cytoskeleton Immunogen sequence: KTAVCDIPPR GLKMSATFIG NNTAIQELFK RVSEQFTAMF RRKAFLHWYT GEGMDEMEFT EAESNMNDLV SEYQQYQDAT AEEEEDEEYA EEEVA.

Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha chain. TUBB8 has a key role in meiotic spindle assembly and oocyte maturation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUBB8
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TUBB8
Gene Accession No. Q3ZCM7
Gene Alias bA631M21.2; HSA10p15 beta-tubulin 4Q; OOMD; OOMD2; RP11-631M21.2; TUBB8; Tubulin beta 8 class VIII; tubulin beta-8 chain
Gene Symbols TUBB8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 350-444 of human TUBB8 (NP_8171241).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347688
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.