Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tubby Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$149.33 - $480.74
Specifications
| Antigen | Tubby |
|---|---|
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB1001922SS
![]() |
Novus Biologicals
NB1001922SS |
0.025 mL |
Each for $149.33
|
|
|||||
NB1001922
![]() |
Novus Biologicals
NB1001922 |
0.1 mL |
Each for $480.74
|
|
|||||
Description
Tubby Polyclonal specifically detects Tubby in C. elegans samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Tubby | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Chromatin Research, Cytoskeleton Markers, Neuroscience, Transcription Factors and Regulators | |
| Whole antisera with 0.05% Sodium Azide | |
| rd5, tubby (mouse) homolog, tubby homolog (mouse), tubby homologue, tubby protein homolog | |
| TUB | |
| IgG | |
| Unpurified | |
| C. elegans. |
| Western Blot 1:1000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| C. elegans | |
| Q09306 | |
| 7275 | |
| Recombinant C. elegans Tubby (domain residues 160-421). [UniProt# Q09306] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title