Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TUBD1 Monoclonal Antibody (3C7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524463
Description
Peptide Sequence: QSADVEGFKD PALYTSWLKP VNAFNVWKTQ RAFSKYEKSA VLVSNSQFLV KPLDMIVGKA WNMFASKAYI HQYTKFGIEE EDFLDSFTSL EQVVASYCNL.
Acts as a positive regulator of hedgehog signaling and regulates ciliary function. [UniProt].
Specifications
TUBD1 | |
Monoclonal | |
Unconjugated | |
TUBD1 | |
4930550G19Rik; Delta-tubulin; Tubd; TUBD1; tubulin delta 1; tubulin delta chain; tubulin, delta 1; tubulin, delta 1-like | |
Mouse | |
Protein A | |
RUO | |
51174 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunoprecipitation | |
3C7 | |
PBS with no preservative; pH 7.4 | |
Q9UJT1 | |
TUBD1 | |
TUBD1 (NP_057345.2, 354 a.a. ~ 453 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction