Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUBD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119485
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119485 200 μL
PIPA5121517 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119485 Supplier Invitrogen™ Supplier No. PA5119485
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TUBD1 Polyclonal Antibody for Western Blot, IHC (P)

Acts as a positive regulator of hedgehog signaling and regulates ciliary function. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUBD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene TUBD1
Gene Accession No. Q9R1K7
Gene Alias 4930550G19Rik; Delta-tubulin; Tubd; TUBD1; tubulin delta 1; tubulin delta chain; tubulin, delta 1; tubulin, delta 1-like
Gene Symbols TUBD1
Host Species Rabbit
Immunogen Recombinant protein Tubulin Delta. The antigen corresponds to amino acid range 236-455 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56427
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.