Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TUBD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $539.50
Specifications
Antigen | TUBD1 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119486
|
Invitrogen™
PA5119486 |
200 μL |
Each for $539.50
|
|
|||||
PIPA5121515
|
Invitrogen™
PA5121515 |
20 μL |
Each for $237.00
|
|
|||||
Description
TUBD1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Acts as a positive regulator of hedgehog signaling and regulates ciliary function. [UniProt].Specifications
TUBD1 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Mouse, Rat | |
TUBD1 | |
4930550G19Rik; Delta-tubulin; Tubd; TUBD1; tubulin delta 1; tubulin delta chain; tubulin, delta 1; tubulin, delta 1-like | |
TUBD1 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q9R1K7 | |
287592, 56427 | |
Recombinant protein Tubulin Delta. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title