Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUBE1 Monoclonal Antibody (5F3B7)
GREENER_CHOICE

Catalog No. PIMA531942
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531942 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531942 Supplier Invitrogen™ Supplier No. MA531942
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31942 has been tested in indirect ELISA.

Microtubules constitute one of the major components of eukaryotic cells cytoskeleton and are involved in many essential processes, including cell division, ciliary and flagellar motility and intracellular transport. Microtubules of the eukaryotic cytoskeleton are composed of a heterodimer of α- and β-tubulin. In addition to α- and β-tubulin, several other tubulins have been identified, bringing the number of distinct tubulin classes to seven. Most of these tubulins have distinct subcellular localization and an emerging, diverse set of functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUBE1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5F3B7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene TUBE1
Gene Accession No. Q9UJT0
Gene Alias 2310061K05Rik; AI551343; dJ142L7.2; epsilon-tubulin; epsilon-tubulin 1; FLJ22589; FLJ44203; RP1-142L7.1; TUBE; Tube1; Tubulin; tubulin epsilon 1; tubulin epsilon chain; tubulin, epsilon 1
Gene Symbols TUBE1
Host Species Mouse
Immunogen Purified recombinant fragment of human TUBE1 (AA: 314-472) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51175
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.