Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TUBG1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144601
Description
TUBG1 Polyclonal Antibody for Western Blot, IHC, IHC (F), IHC (P)
This gene encodes a member of the tubulin superfamily. The encoded protein localizes to the centrosome where it binds to microtubules as part of a complex referred to as the gamma-tubulin ring complex. The protein mediates microtubule nucleation and is required for microtubule formation and progression of the cell cycle. A pseudogene of this gene is found on chromosome 7.
Specifications
TUBG1 | |
Polyclonal | |
Unconjugated | |
TUBG1 | |
TUBG1 | |
The immunogen used to generate this antibody corresponds to zebrafish Tubg1. | |
100 μL | |
Primary | |
Zebrafish | |
Antibody | |
IgG |
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal | |
1500010O08Rik; AI451582; AI503389; CDCBM4; gamma-1-tubulin; Gamma-tubulin complex component 1; GCP-1; Tubg; TUBG1; TUBGCP1; tubulin gamma 1; tubulin gamma-1 chain; tubulin, gamma 1; tubulin, gamma polypeptide | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
393882 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction