Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBG1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA589901
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589901 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589901 Supplier Thermo Scientific Supplier No. PA589901

Rabbit Polyclonal Antibody

Immunogen sequence: PWGPASIQVA LSRKSPYLPS AHRVSGLMMA NHTSISSLFE RTCRQYDKLR KREAFLEQFR KEDMFKDNFD EMDTSREIVQ QLIDEYHAAT RPDYISWGTQ E; Positive Samples: MCF-7, HeLa, Mouse brain, Mouse lung, Rat brain; Cellular Location: Cytoplasm, centrosome, cytoskeleton, microtubule organizing center

This gene encodes a member of the tubulin superfamily. The encoded protein localizes to the centrosome where it binds to microtubules as part of a complex referred to as the gamma-tubulin ring complex. The protein mediates microtubule nucleation and is required for microtubule formation and progression of the cell cycle. A pseudogene of this gene is found on chromosome 7.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUBG1
Applications Western Blot
Classification Polyclonal
Concentration 2.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal, pH 7.3
Gene TUBG1
Gene Accession No. P23258, P83887, P83888
Gene Alias 1500010O08Rik, AI451582, AI503389, CDCBM4, gamma-1-tubulin, Gamma-tubulin complex component 1, GCP-1, Tubg, TUBG1, TUBGCP1, tubulin gamma 1, tubulin gamma-1 chain, tubulin, gamma 1, tubulin, gamma polypeptide
Gene Symbols TUBG1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 350-450 of human γ-Tubulin (NP_0010612)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103733, 252921, 7283
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.