Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUBG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113396
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113396 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113396 Supplier Invitrogen™ Supplier No. PA5113396
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TUBG2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Tubulin is the major constituent of microtubules. The gamma chain is found at microtubule organizing centers (MTOC) such as the spindle poles or the centrosome. Pericentriolar matrix component that regulates alpha/beta chain minus-end nucleation, centrosome duplication and spindle formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUBG2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Tubg2
Gene Accession No. Q8VCK3, Q9NRH3
Gene Alias AI504772; Gamma-2-tubulin; Tubg2; Tubgl; tubulin gamma 2; tubulin gamma-2 chain; tubulin, gamma 2; Unknown (protein for MGC:133882)
Gene Symbols Tubg2
Host Species Rabbit
Immunogen Recombinant Human Tubulin gamma-2 chain protein (276-451AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103768, 27175
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.