Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TUBGCP2 Antibody (GCP2-01), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP221793AF405
Description
TUBGCP2 Monoclonal specifically detects TUBGCP2 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TUBGCP2 | |
Monoclonal | |
Alexa Fluor 405 | |
Gamma-ring complex protein 103 kDa, gamma-tubulin complex component 2, gamma-tubulin complex protein 2, GCP-2, GCP2hGrip103, h103p, hGCP2, hSpc97, MGC138162, SPBC97, Spc97p, Spindle pole body protein Spc97 homolog, tubulin, gamma complex associated protein 2 | |
Mouse | |
Protein A purified | |
RUO | |
10844 | |
Human, Mouse, Rat, Pig | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
GCP2-01 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
TUBGCP2 | |
GST-fusion protein containing amino acids 2-194 of mouse GCP2. | |
0.1 mL | |
Primary | |
The mouse monoclonal antibody GCP2-01 recognizes gamma-tubulin complex component 2 (GCP2), a 95-103 kDa protein. It recognizes also phosphorylated GCP2. | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction