Learn More
Bel-Art™ SP Scienceware™ Metric I.D./O.D. Gauge
SCI_ED
Catalog No. S01701 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Click to view available options
Material:
Polypropylene
Polypropylene metric diameter gauge for tubing and rods
Handy tool for measuring exact size of metric tubing — keep one gauge with tubing supplies and one with labware.
  • Polypropylene tool measures glass, plastic or rubber tubing, as well as connectors, rods and test tubes Post side measures I.D.; reverse side measures O.D.
  • Fits range of sizes from 0.05 to 0.63 in. (1.5 to 16mm)
  • Autoclavable
Specifications
Autoclavable Autoclavable
Material Polypropylene
For Use With Plastic, rubber tubing, connecting pieces, rods, test tubes
Product Type Metric Diameter Gauge Tubing Sizer
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.