Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tubulin alpha-1B Antibody (OTI3G3), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274703IR
Description
Tubulin alpha-1B Monoclonal specifically detects Tubulin alpha-1B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Tubulin alpha-1B | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TUBA1B | |
Full length recombinant protein of human Tubulin alpha-1B (NP_006073) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI3G3 | |
Western Blot, Immunohistochemistry | |
alpha tubulin, Alpha-tubulin ubiquitous, K-ALPHA-1, tubulin alpha-1B chain, Tubulin alpha-ubiquitous chain, Tubulin K-alpha-1, tubulin, alpha 1b, tubulin, alpha, ubiquitous | |
Mouse | |
Affinity Purified | |
Cell Biology, Cytoskeleton Markers, Stem Cell Markers | |
10376.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction