Learn More
Invitrogen™ Tubulin alpha-1B Monoclonal Antibody (OTI3G3)

Description
Tubulin alpha-1B Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | Tubulin alpha-1B |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI3G3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | TUBA1B |
| Gene Accession No. | P68363 |
| Gene Alias | alpha tubulin; alpha-tubulin 2; Alpha-tubulin isotype M-alpha-2; alpha-tubulin ubiquitous; Detyrosinated tubulin alpha-1B chain; K-ALPHA-1; RGD1565476; Tuba1b; Tuba2; tubulin alpha 1b; tubulin alpha-1B chain; Tubulin alpha-2 chain; tubulin alpha-ubiquitous chain; Tubulin K-alpha-1; tubulin, alpha 1B; tubulin, alpha 2; tubulin, alpha, ubiquitous |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.