Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tubulin alpha-3E Antibody (OTI2A8), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274704MFV610
Description
Tubulin alpha-3E Monoclonal antibody specifically detects Tubulin alpha-3E in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
| Tubulin alpha-3E | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 112714 | |
| Store at 4C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| OTI2A8 | |
| Flow Cytometry | |
| Alpha-tubulin 3E, tubulin alpha-3E chain, tubulin, alpha 3e | |
| Full length human recombinant protein of human TUBA3E (NP_997195) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction