Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tubulin alpha-3E Antibody (OTI2A8), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274704PE
Description
Tubulin alpha-3E Monoclonal specifically detects Tubulin alpha-3E in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
Tubulin alpha-3E | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
TUBA3E | |
Full length recombinant protein of human Tubulin alpha-3E (NP_997195) produced in HEK293T cells. | |
0.1 mL | |
112714.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI2A8 | |
Flow Cytometry | |
Alpha-tubulin 3E, tubulin alpha-3E chain, tubulin, alpha 3e | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction