Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tubulin-alpha Polyclonal Antibody, Bioss
SDP

Catalog No. 502280308
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-0308 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-0308 Supplier Bioss Supplier No. BS0159R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Tubulin-alpha Polyclonal antibody specifically detects Tubulin-alpha in Human, Mouse, Plant, Rat samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Tubulin-alpha
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene TUBA1C
Gene Accession No. P05213, P0DPH7, P68363, P68366, P68368, P68369, P68370, P68373, Q5XIF6, Q6AYZ1, Q6P9V9, Q6PEY2, Q71U36, Q9BQE3
Gene Alias &agr-tubulin 84B; 1t; a1t; ALPHA 84C; alpha tubulin; alpha tubulin 84B; alpha[[1]] tubulin; alpha[[1]]-tubulin; alpha1; alpha1 tubulin; alpha1t; alpha1tub; alpha1tub84B; alpha1-tubulin; alpha84B; alphaT; alphat-1; alphatub; alpha-tub; alphaTub1; alphatub84; alphaTUB84B; alpha-tub84B; alphaTub84B-PA; alphatubulin; alpha-tubulin; Alpha-tubulin 1; Alpha-tubulin 2; Alpha-tubulin 3; Alpha-tubulin 3C; alpha-tubulin 3C/D; Alpha-tubulin 3D; Alpha-tubulin 3E; alpha-tubulin 4; Alpha-tubulin 6; alphaTubulin 84B; alpha-Tubulin 84B; alpha-Tubulin at 84B; alpha-tubulin I; Alpha-tubulin II; alpha-tubulin III; Alpha-tubulin isotype M-alpha-1; alpha-tubulin isotype M-alpha-2; Alpha-tubulin isotype M-alpha-4; alpha-tubulin isotype M-alpha-6; alpha-tubulin TUB1; alpha-tubulin ubiquitous; alphaTubulin84B; Alpha-Tubulin84B; ALS22; anon-EST:Liang-1.59; anon-EST:Liang-2.30; AT25469p; aTub84B; a-tub84B; bA408E5.3; B-ALPHA-1; bcm948; BEST:LD32507; cb944; CG1913; CG1913-PA; chr3R:2914276..2914416; clone 1.59; c
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Tubulin-alpha 1 (amino acids 375-448).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10376, 112714, 22142, 22143, 22145, 22146, 300218, 316531, 500929, 64158, 7277, 7278, 7846, 84790
Target Species Human, Mouse, Plant, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.