Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tubulin beta-2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530379
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530379 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530379 Supplier Invitrogen™ Supplier No. PA530379
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat, Raji, NIH-3T3. Predicted reactivity: Mouse (100%), Rat (99%), Zebrafish (100%), Xenopus laevis (100%), Chicken (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha-chain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tubulin beta-2C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene TUBB4B
Gene Accession No. P68371, P68372, Q6P9T8
Gene Alias 4930542G03Rik; Beta2; class IVb beta tubulin; class IVb beta-tubulin; LOW QUALITY PROTEIN: tubulin beta chain; novel tubulin protein; Tubb2; TUBB2C; Tubb2c1; TUBB4B; tubulin beta; tubulin beta 4B class IVb; tubulin beta-2 chain; Tubulin beta-2C chain; tubulin beta-4B chain; tubulin, beta 2C; tubulin, beta 2c1; tubulin, beta 4B class IVb; tubulin, beta, 2
Gene Symbols TUBB4B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human beta Tubulin 2C. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10383, 227613, 296554
Target Species Human, Mouse, Monkey, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.