Learn More
Invitrogen™ Tubulin beta-2C Polyclonal Antibody

Description
This antibody is predicted to react with bovine, chicken, canine, hamster, mouse, porcine, rat and Xenopus based on sequence homology.
Specifications
Specifications
| Antigen | Tubulin beta-2C |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | TUBB4B |
| Gene Accession No. | P68371 |
| Gene Alias | 4930542G03Rik; Beta2; class IVb beta tubulin; class IVb beta-tubulin; LOW QUALITY PROTEIN: tubulin beta chain; novel tubulin protein; Tubb2; TUBB2C; Tubb2c1; TUBB4B; tubulin beta; tubulin beta 4B class IVb; tubulin beta-2 chain; Tubulin beta-2C chain; tubulin beta-4B chain; tubulin, beta 2C; tubulin, beta 2c1; tubulin, beta 4B class IVb; tubulin, beta, 2 |
| Gene Symbols | TUBB4B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.