Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tubulin Beta 4 Antibody (OTI3F1), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274713AF594
Description
Tubulin Beta 4 Monoclonal specifically detects Tubulin Beta 4 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Tubulin Beta 4 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
TUBB4A | |
Full length recombinant protein of human Tubulin Beta 4 (NP_006078) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3F1 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
TUBB5beta-5, Tubulin 5 beta, tubulin beta-4 chain, tubulin, beta 4, tubulin, beta, 5 | |
Mouse | |
Affinity Purified | |
Apoptosis, Cytoskeleton Markers | |
10382.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction