Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tubulin Beta 4 Antibody (OTI3F1), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274713PCP
Description
Tubulin Beta 4 Monoclonal specifically detects Tubulin Beta 4 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.Specifications
| Tubulin Beta 4 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| TUBB4A | |
| Full length recombinant protein of human Tubulin Beta 4 (NP_006078) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG2a |
| Flow Cytometry | |
| OTI3F1 | |
| Flow Cytometry | |
| TUBB5beta-5, Tubulin 5 beta, tubulin beta-4 chain, tubulin, beta 4, tubulin, beta, 5 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Cytoskeleton Markers | |
| 10382.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction