Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tubulin beta class V Mouse anti-Canine, Human, Mouse, Clone: 1D7B9, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Tubulin beta class V |
---|---|
Clone | 1D7B9 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6792
![]() ![]() |
Proteintech Group Inc
663621IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha chain.Specifications
Tubulin beta class V | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HsT1601, TUBB 5, TUBB6, Tubulin beta 6 chain, Tubulin beta class V, tubulin, beta 6 | |
TUBB6 | |
IgG1 | |
Protein G | |
Antibody |
1D7B9 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Canine, Human, Mouse | |
Q922F4, Q9BUF5 | |
480213, 67951, 84617 | |
Peptide | |
Primary | |
-20°C | |
TUBB6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title