Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ α-Tubulin, Mouse, Alexa Fluor 555, Clone: DM1A,

Catalog No. 05829AF555 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 555
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
05-829-AF555 100 μL Alexa Fluor 555
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-829-AF555 Supplier MilliporeSigma™ Supplier No. 05829AF555
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Voltage-dependent L-type calcium channel subunit alpha-1S (UniProt P07293; also known as Calcium channel, L type, alpha-1 polypeptide, isoform 3, skeletal muscle, Voltage-gated calcium channel subunit alpha Cav1.1) is encoded by the CACNA1S (also known as CACH1, CACN1, CACNL1A3) gene (Gene ID 100009585) in Oryctolagus cuniculus (rabbit) species. Voltage-dependent calcium channels (VDCC) are found in the membrane of excitable cells (e.g., muscle, glial cells, neurons) and exhibit 1000-fold greater permeability to calcium than to sodium under normal physiological conditions. There exist five types of VDCCs, including four high voltage-activated channels (Neural- or N-type channel, Residual- or R-type channel, Purkinje- or P/Q-type channel, and the dihydropyridine-sensitive Long-lasting or L-type channels) and the low voltage-activated Transient or T-type calcium channels. VDCCs exist as a complex of different subunits (α1, α2δ, β1-4, and γ), with α1 being the the ion-conducting pore-forming subunit that contains voltage-sensing machinery and the drug/toxin-binding sites. There exist ten genes coding for different α1 subunits, four of which code for L-type alpha-1 subunits (alpha-1A/Cav2.1/CACNA1A, alpha-1C/Cav1.2/CACNA1C, alpha-1D/Cav1.3/CACNA1D, and alpha-1S/Cav1.1/CACNA1S). The dihydropyridine-sensitive L-type channels are responsible for excitation-contraction coupling of skeletal, smooth, and cardiac muscle and for hormone secretion in endocrine cells. The α1S subunit (Cav1.1) contains the characteristic four homologous I–IV domains (a.a. 38-337, 418-664, 786-1068, 1105-1384) with six transmembrane α-helices each, having both its N- and C-terminal ends exposed intracellularly (a.a. 1-51, 1382-1873).
TRUSTED_SUSTAINABILITY

Specifications

Antigen α-Tubulin
Applications Immunocytochemistry
Classification Monoclonal
Clone DM1A
Concentration Please refer to lot specific datasheet.
Conjugate Alexa Fluor 555
Formulation Purified mouse monoclonal IgG1 antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % Sodium Azide.
Gene TUBA1C, TUBA1
Gene Accession No. P02552
Host Species Mouse
Immunogen Purified chicken brain α-tubulin.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001292201.1
Test Specificity Clone DM1A recognizes α-Tubulin, MW 50-60 kDa. The epitope recognized by this antibody has been mapped to the C-terminal 25 amino acids (AALEKDYEEVGVDSVEGEGEEEGEE).
Target Species Avian
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.