Learn More
MP Biomedicals™ Tubulin, Purified from Bovine Brain
Tubulin, Purified from Bovine Brain is a heterodimer composed of alpha- and beta-tubulin of 55 kDa (molecular weight ∼110 kDa).
Supplier:  MP Biomedicals™ 08771121
Catalog No. 08-771-121
Only null left
Add to Cart
Description
Description
  • Purified bovine brain tubulin protein supplied in liquid form at 10 mg/mL in 80 mM PIPES buffer containing 1 mM EGTA, 1 mM GTP, pH 6.8 and 10% glycerol.
  • It is a heterodimer composed of α- and β- tubulin of 55 kDa (molecular weight ≅110 kDa).
  • Polymerization occurs forming microtubules of 25 nm in diameter and average 1650 heterodimers per μm in length.
  • One unit of tubulin is defined as 5.0 mg of purified protein (as determined by the method of Bradford: Bradford, M., Analytical Biochemistry 72, 248, 1976).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Preparation Method
One unit of tubulin in G-PEM buffer pH 6.8 plus 10% glycerol was incubated at 35°C in a cuvette. The optical density at 340 nm was taken at time intervals. A value of 1.0 OD unit indicates that >97% of the tubulin has polymerized. The polymerization reaction was performed in duplicate.
1 mg
When tubulin at a concentration of 1.0 unit/mL (in G-PEM buffer plus 10% glycerol [v/v]) is incubated at 35°C for 30 minutes an increase in optical density (OD340nm) from 0.0 to 1.0 is observed. This corresponds to a total microtubule (MT) polymer mass of >4.8 mg/mL. The mean length of the microtubule population under these conditions is approximately 10 μm as measured by immunoflourescence microscopy and the average number of MTs per ml is 1.0 x 1012.
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.