Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tucatinib
GREENER_CHOICESDP

Catalog No. 764050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-405-0 50 mg
76-401-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 76-405-0 Supplier Tocris Bioscience™ Supplier No. 7640/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent and selective ErbB2 inhibitor

Tucatinib is a potent and selective ErbB-2 (also called HER2) kinase inhibitor (IC50 = 7 nM). It exhibits >50-fold selectivity for ErbB-2 over EGFR. In vitro, Tucatinib inhibits ErbB-2 signaling activity and cell proliferation in ErbB-2-amplified breast cancer cell lines (BT-474). In vivo, Tucatinib (25-100 mg/kg) reduces tumor volume in breast and gastric xenograft mouse models.This compound is orally bioavailable.

Specifications

Chemical Name or Material N6-(4,5-Dihydro-4,4-dimethyl-2-oxazolyl)-N4-[3-methyl-4-([1,2,4 ]triazolo[1,5-a]pyridin-7-yloxy)phenyl]-4,6-quinazolinediamine
CAS 937263-43-9
Quantity 50 mg
Target EGF Receptor Inhibitors
Molecular Formula C26H24N8O2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.