Learn More
Bio-Serv™ Tuff Turf Foraging Boards
$29.20
Catalog Number | Mfr. No. | Size | Dimensions (L x W) | Material | Product Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Dimensions (L x W) | Material | Product Type | Price | Quantity & Availability | ||
14-727-037
![]() ![]() |
Bio-Serv™
F9114R |
Large | 15.5 x 6 in. | Artificial Turf | Replacement Pad |
Each of 1 for $29.20
|
|
||
Description
- Made of Stainless Steel
- Available in 2 sizes
- Extra-durable artificial turf is securely anchored by stainless steel bars and lock nuts
- Texture of turf hides small treats increasing foraging time
- Tray has drainage holes for easy cleaning
- Replacement turf pads available
- Autoclavable (Board Only)
- Cagewasher safe