Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Maximum Safety™ TuffMax3™ HPPE Gloves, Nitrile Micro-Surface Palm

Catalog No. 19152938 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-152-938 Large
19-152-937 Medium
19-152-939 X-Large
19-152-940 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-152-938 Supplier PIP™ Supplier No. 1205130/L
Add to Cart
Add to Cart
This item is not returnable. View return policy

High performance polyethylene (HPPE) fiber shell is lightweight and provides dexterity, tactile sensitivity and cut resistance. Ideal for petrochemical, construction, auto repair, demo, mining and fabrication industries.

  • 13-gauge high performance polyethylene (HPPE) fiber shell is light-weight and provides dexterity, tactile sensitivity and cut resistance
  • Micro-surface nitrile-coated padded palm for enhanced wet and dry grip
  • Thermoplastic rubber (TPR) molded finger, thumb and back of hand guards for maximum protection
  • Reinforced leather between thumb and index finger
  • Extended wrist
  • Hook and loop closure for easy on/off and a secure fit

Specifications

ANSI Abrasion Rating ANSI 2
Certifications/Compliance ASTM F2992-15 ANSI Cut Level A2 (Adopted by ANSI/ISEA 02/16)/ASTM F1790-05 ANSI Cut Level 2
Coating Material MicroSurface Nitrile
Color Lime Green, Black
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free Yes
Material HPPE, Thermoplastic Rubber
Product Type Cut-resistant Gloves
Size Large
EN388 Standard EN 388:4343
For Use With (Application) Petrochemical, construction, auto repair, demolition, mining, fabrication
Model TDR impact protection
Series Maximum Safety™ TuffMax3™
Thumb Style Reinforced
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.