Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TUFM Antibody (900), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311261
Description
TUFM Monoclonal specifically detects TUFM in E. coli samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoassay.Specifications
TUFM | |
Monoclonal | |
0.1 mg/ml | |
Western Blot, Immunocytochemistry/Immunofluorescence, Immunoassay | |
EFTu, EF-Tu, EFTUCOXPD4, EF-TuMT, elongation factor Tu, mitochondrial, P43, Tu translation elongation factor, mitochondrial | |
E. coli K-12 C600 Rif ( pKT146) bacteria | |
50 μg | |
Primary | |
E. coli | |
Purified |
Western Blot, Immunofluorescence, Immunoassay | |
900 | |
Unconjugated | |
0.2 um filtered solution in PBS, 0.1% BSA | |
Mouse | |
Protein G purified | |
RUO | |
7284 | |
Store at 4°C. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction