Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TUFM Antibody (CL2245), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | TUFM |
---|---|
Clone | CL2245 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
TUFM Monoclonal specifically detects TUFM in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TUFM | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Human | |
P49411 | |
7284 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:AVDTYIPVPARDLEKPFLLPVEAVYSVPGRGTVVTGTLERGILKKGDECELLGHSKNIRTVVTGIEMFHKSLERAEAGDNLGALVRGLKREDLRRGLVMVKPGSI | |
Primary | |
Protein A purified |
CL2245 | |
Monoclonal | |
Purified | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
EFTu, EF-Tu, EFTUCOXPD4, EF-TuMT, elongation factor Tu, mitochondrial, P43, Tu translation elongation factor, mitochondrial | |
TUFM | |
IgG1 | |
Protein A purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title