Learn More
TUFM/EF-Tu Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305014AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine and Golden hamster
This gene encodes a protein which participates in protein translation in mitochondria. Mutations in this gene have been associated with combined oxidative phosphorylation deficiency resulting in lactic acidosis and fatal encephalopathy. A pseudogene has been identified on chromosome 17.Specifications
TUFM/EF-Tu | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P49411, Q8BFR5 | |
TUFM | |
Between 100 and 150 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
TUFM | |
COXPD4, EF-TuMT, EFTU, P43, EF-Tu, elongation factor Tu, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
233870, 7284 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.