Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUFM Monoclonal Antibody (CL2245)
GREENER_CHOICE

Catalog No. PIMA534990
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534990 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534990 Supplier Invitrogen™ Supplier No. MA534990
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: AVDTYIPVPA RDLEKPFLLP VEAVYSVPGR GTVVTGTLER GILKKGDECE LLGHSKNIRT VVTGIEMFHK SLERAEAGDN LGALVRGLKR EDLRRGLVMV KPGSI Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 95%.

This gene encodes a protein which participates in protein translation in mitochondria. Mutations in this gene have been associated with combined oxidative phosphorylation deficiency resulting in lactic acidosis and fatal encephalopathy. A pseudogene has been identified on chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUFM
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone CL2245
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TUFM
Gene Accession No. P49411
Gene Alias 2300002G02Rik; C76308; C76389; COXPD4; D250; EFTU; EF-Tu; EF-TuMT; elongation factor Tu; elongation factor Tu, mitochondrial; fi06f04; P43; Tu translation elongation factor, mitochondrial; TUFM; tufm {ECO:0000250; UniProtKB:P49411}; wu:fi06f04; zgc:110766
Gene Symbols TUFM
Host Species Mouse
Immunogen Recombinant protein fragment of Human TUFM (Product #RP-92517)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7284
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.