Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUFM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544906
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544906 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544906 Supplier Invitrogen™ Supplier No. PA544906
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PEKELAMPGE DLKFNLILRQ PMILEKGQRF TLRDGNRTIG TGLVTNTLAM Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 93%; Yeast: 77%; Zebrafish: 86%.

This gene encodes a protein which participates in protein translation in mitochondria. Mutations in this gene have been associated with combined oxidative phosphorylation deficiency resulting in lactic acidosis and fatal encephalopathy. A pseudogene has been identified on chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUFM
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TUFM
Gene Accession No. P49411
Gene Alias 2300002G02Rik; C76308; C76389; COXPD4; D250; EFTU; EF-Tu; EF-TuMT; elongation factor Tu; elongation factor Tu, mitochondrial; fi06f04; P43; Tu translation elongation factor, mitochondrial; TUFM; tufm {ECO:0000250; UniProtKB:P49411}; wu:fi06f04; zgc:110766
Gene Symbols TUFM
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TUFM.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7284
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.